Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cathepsin Z Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119013
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119013 200 μL
PIPA5122153 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119013 Supplier Invitrogen™ Supplier No. PA5119013
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Cathepsin Z Polyclonal Antibody for Western Blot, ICC/IF, IHC

The protein encoded by this gene is a lysosomal cysteine proteinase and member of the peptidase C1 family. It exhibits both carboxy-monopeptidase and carboxy-dipeptidase activities. The encoded protein has also been known as cathepsin X and cathepsin P. This gene is expressed ubiquitously in cancer cell lines and primary tumors and, like other members of this family, may be involved in tumorigenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathepsin Z
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Ctsz
Gene Accession No. Q9R1T3, Q9UBR2
Gene Alias AI787083; AU019819; carboxypeptidase LB; cathepsin B2; cathepsin IV; cathepsin P; Cathepsin X; cathepsin Y; cathepsin Z; cathepsin Z1; CTSX; CTSZ; cysteine-type carboxypeptidase; D2Wsu143e; lysosomal carboxypeptidase B; preprocathepsin P
Gene Symbols Ctsz
Host Species Rabbit
Immunogen Recombinant protein Cathepsin Z. The antigen corresponds to amino acid range 24-303 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1522, 252929
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.