Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cathepsin Z Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562072
Description
Immunogen sequence: PDETCNNYQA KDQECDKFNQ CGTCNEFKEC HAIRNYTLWR VGDYGSLSGR EKMMAEIYAN GPISCGIMAT ERLANYTGGI YAEYQD Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.
The protein encoded by this gene is a lysosomal cysteine proteinase and member of the peptidase C1 family. It exhibits both carboxy-monopeptidase and carboxy-dipeptidase activities. The encoded protein has also been known as cathepsin X and cathepsin P. This gene is expressed ubiquitously in cancer cell lines and primary tumors and, like other members of this family, may be involved in tumorigenesis.
Specifications
Cathepsin Z | |
Polyclonal | |
Unconjugated | |
Ctsz | |
AI787083; AU019819; carboxypeptidase LB; cathepsin B2; cathepsin IV; cathepsin P; Cathepsin X; cathepsin Y; cathepsin Z; cathepsin Z1; CTSX; CTSZ; cysteine-type carboxypeptidase; D2Wsu143e; lysosomal carboxypeptidase B; preprocathepsin P | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1522 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UBR2 | |
Ctsz | |
Recombinant protein corresponding to Human Cathepsin Z. Recombinant protein control fragment (Product #RP-103060). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction