Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cathepsin Z Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580523
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580523 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580523 Supplier Invitrogen™ Supplier No. PA580523
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human CTSZ/CTSX/Cathepsin Z.

The protein encoded by this gene is a lysosomal cysteine proteinase and member of the peptidase C1 family. It exhibits both carboxy-monopeptidase and carboxy-dipeptidase activities. The encoded protein has also been known as cathepsin X and cathepsin P. This gene is expressed ubiquitously in cancer cell lines and primary tumors and, like other members of this family, may be involved in tumorigenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathepsin Z
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Ctsz
Gene Accession No. Q9UBR2
Gene Alias AI787083; AU019819; carboxypeptidase LB; cathepsin B2; cathepsin IV; cathepsin P; Cathepsin X; cathepsin Y; cathepsin Z; cathepsin Z1; CTSX; CTSZ; cysteine-type carboxypeptidase; D2Wsu143e; lysosomal carboxypeptidase B; preprocathepsin P
Gene Symbols Ctsz
Host Species Rabbit
Immunogen Recombinant Human CTSZ/CTSX/Cathepsin Z protein (Met1-Val303).
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1522
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.