Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin Z Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABC442
Description
This Anti-Cathepsin Z antibody is validated for use in Western Blotting for the detection of Cathepsin Z.
Specifications
Cathepsin Z | |
Polyclonal | |
Unconjugated | |
Cathepsin Z;Cathepsin P;Cathepsin X | |
Rabbit | |
∽33 kDa observed. (Pungecar, J., et al. (2000). PMID: 10653163) | |
100 μg | |
Apoptosis & Cancer | |
NP_001327 | |
Stable for 1 year at 2-8°C from date of receipt. |
Western Blot | |
Please refer to lot specific datasheet. | |
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
CTSZ | |
KLH-conjugated linear peptide corresponding to human Cathepsin Z near the C-terminus. | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibodies |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction