Learn More
3B Scientific™ Catheterization simulator PRO
GREENER_CHOICESDPSCI_ED
Catalog No. p-7221827 Shop All 3B Scientific Products
Change view
Click to view available options
Gender:
Female
Male
Set
Catalog No. Gender
S58102 Female
S58103 Male
S58104 Set
3 options
Catalog No. S58102
Supplier: 3B Scientific™ 1023008
Only null left
Add to Cart
Add to Cart
Used to realistically demonstrate, practice and evaluate female bladder catheterizations either via the urethra (transurethral) or the abdominal wall (suprapubic).
  • The genital inserts are placed into the anatomically correct lower abdominal model and held in place with magnets. The material is soft and flexible: the labia are movable and can be separated.
  • This way the student can practice all the necessary gestures for catheterization (e.g. disinfection). The material gives a realistic feeling when inserting and removing the catheter into the bladder with realistic resistance and insertion depths.
  • Catheterization with realistic resistance
  • Soft and movable (for practicing disinfection, for example)
  • Anatomically realistic pelvic structure
  • Liquid outflow if catheterization is successfully carried out
  • The transparent bladder can be checked, and the abdominal wall can be removed
  • Training of suprapubic catheter changes as well as their cleaning and care
  • Stands securely on the table thanks to the non-slip feet
  • Magnetic connectors for quick set-up and dismantling
  • Easy to clean and maintain, and can be completely taken apart
  • A sponge on the inside prevents a build-up of moisture
Specifications
Product Type Catheterization Trainer
For Use With (Application) Female Catheterization Training
Gender Female