Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CATIP Antibody (OTI4B4), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272506G
Description
CATIP Monoclonal specifically detects CATIP in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
CATIP | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CATIP | |
Full length recombinant protein of human C2orf62 (NP_940961) produced in HEK293T cells. | |
0.1 mL | |
375307.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI4B4 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
C2orf62, chromosome 2 open reading frame 62, hypothetical protein LOC375307, MGC50811 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction