Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CATIP Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544446
Description
Peptide sequence: MSSKVYSTGS RAKDHQPSGP ECLPLPEANA EAIDFLSSLH KEELQMLFFS Sequence homology: Human: 100%.
Plays a role in primary ciliogenesis by modulating actin polymerization.
Specifications
| CATIP | |
| Polyclonal | |
| Unconjugated | |
| CATIP | |
| C15H2orf62; C2orf62; C37H2orf62; C6H2orf62; Catip; ciliogenesis associated TTC17 interacting protein; Ciliogenesis-associated TTC17-interacting protein; Gm216; hypothetical protein LOC690437; LOC690437 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 375307 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q7Z7H3 | |
| CATIP | |
| Synthetic peptide directed towards the N-terminal of human C2orf62 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction