Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CATPB
GREENER_CHOICESDP

Catalog No. 590310 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-031-0 10 mg
59-035-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 59-031-0 Supplier Tocris Bioscience™ Supplier No. 5903/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

FFA2 antagonist/inverse agonist

FFA2 inverse agonist (pKi = 7.87). Selective for human over mouse FFA2. Increases forskolin-induced cAMP production and inhibits acetate-induced MAPK signaling in cells expressing human FFA2.

Chemical Identifiers

CAS 1322598-09-3
Molecular Formula C19H17ClF3NO3

Specifications

Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 50mM in ethanol
Formula Weight 399.79
Physical Form Solid
Purity >98%
Molecular Weight (g/mol) 399.794
Chemical Name or Material (S)-3-(2-(3-Chlorophenyl)acetamido)-4-(4-(trifluoromethyl)phenyl)butanoic acid
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.