Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CATSPER1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5103715

Catalog No. PIPA5103715


Only null left
Add to Cart

Description

Description

Antibody detects endogenous levels of total CATSPER1.

Voltage-gated calcium channel that plays a central role in calcium-dependent physiological responses essential for successful fertilization, such as sperm hyperactivation, acrosome reaction and chemotaxis towards the oocyte. Activated by extracellular progesterone and prostaglandins following the sequence: progesterone > PGF1-alpha = PGE1 > PGA1 > PGE2 >> PGD2. The primary effect of progesterone activation is to shift voltage dependence towards more physiological, negative membrane potentials; it is not mediated by metabotropic receptors and second messengers. Sperm capacitation enhances the effect of progesterone by providing additional negative shift. Also activated by the elevation of intracellular pH.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CATSPER1
Polyclonal
Unconjugated
CATSPER1
cation channel of sperm 1; cation channel sperm associated 1; cation channel sperm-associated protein 1; cation channel, sperm associated 1; CATSPER; CATSPER1; hCatSper; KSper; sperm ion channel; sperm protein CATSPER1; sperm-associated cation channel 1; SPGF7
Rabbit
Affinity chromatography
RUO
117144, 225865
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q8NEC5, Q91ZR5
CATSPER1
A synthesized peptide derived from human CATSPER1(Accession Q8NEC5), corresponding to amino acid residues P30-H80.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.