Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CATSPER2 Polyclonal Antibody

CATSPER2 Polyclonal Antibody
Supplier: Thermo Scientific PA598703
Description
CATSPER2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Specifications
CATSPER2 | |
Polyclonal | |
Unconjugated | |
CATSPER2 | |
cation channel sperm associated 2; cation channel sperm-associated protein 2; cation channel, sperm associated 2; CATSPER2; sperm ion channel; sperm-associated cation channel 2 | |
Rabbit | |
Protein G | |
RUO | |
117155 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
4 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q96P56 | |
CATSPER2 | |
Recombinant Human Cation channel sperm-associated protein 2 protein (5-112AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction