Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CATSPER3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103716
Description
Antibody detects endogenous levels of total CATSPER3.
CatSpers (cation channel, sperm associated proteins) are ion transport proteins located on the surface of sperm cells in the principal piece of the sperm tail. CatSpers are vital to sperm motility, fertilization and cAMP-mediated calcium influx in sperm. There are four CatSper proteins in mammalian sperm, namely CatSper (or CatSper1), CatSper2, CatSper3 and CatSper4. CatSper proteins contain a single, six-transmembrane-spanning segment and exhibit the voltage-dependent Ca2+ channel four-repeat structure. CatSper proteins are believed to assemble into a heterotetrameric complex, forming an alkalinization-activated Ca2+-selective channel. Mutations in any of the genes encoding CatSper family proteins can result in male infertility. CatSper3 plays an important role in the hyperactivated motility of sperm cells, a process that is required in the preparation of sperm for fertilization.
Specifications
CATSPER3 | |
Polyclonal | |
Unconjugated | |
CATSPER3 | |
4921522D01Rik; Ca(v)-like protein; CACRC; calcium channel repeat containing 1; cation channel sperm associated 3; cation channel sperm-associated protein 3; cation channel, sperm associated 3; CatSper3; One-repeat calcium channel-like protein; putative ion channel CatSper3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
347732, 76856 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q80W99, Q86XQ3 | |
CATSPER3 | |
A synthesized peptide derived from human CATSPER3(Accession Q86XQ3), corresponding to amino acid residues L183-Q233. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction