Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CATSPER4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119864
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119864 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119864 Supplier Invitrogen™ Supplier No. PA5119864
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: AGS cell lysates, MG-63. Subcellular Location: Flagellum membrane.

CatSper4, is a protein which in humans is encoded by the CATSPER4 gene. CatSper1 is a member of thecation channels of sperm family of protein. The four proteins in this family together form a Ca2+-permeant ion channel specific essential for the correct function of sperm cells. Voltage-gated calcium channel that plays a central role in calcium-dependent physiological responses essential for successful fertilization, such as sperm hyperactivation, acrosome reaction and chemotaxis towards the oocyte.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CATSPER4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene CATSPER4
Gene Accession No. Q7RTX7
Gene Alias 4933412L05; cation channel of sperm 4; cation channel sperm associated 4; cation channel sperm-associated protein 4; cation channel, sperm associated 4; CatSper4; channel, sperm associated 4; CTSR4; ion channel CatSper4; RGD1565213
Gene Symbols CATSPER4
Host Species Rabbit
Immunogen Recombinant protein within human CATSPER4 aa 306-472.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 378807
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.