Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CATSPER4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560965
Description
Immunogen sequence: GQQQRITFSE TGAEEEEEND QLPLVHCVVA RSEKSGLLQE PLAGGPLSNL SENTCDNFCL VLEAIQENLR QYKEIRD Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 71%.
CatSper4, is a protein which in humans is encoded by the CATSPER4 gene. CatSper1 is a member of thecation channels of sperm family of protein. The four proteins in this family together form a Ca2+-permeant ion channel specific essential for the correct function of sperm cells. Voltage-gated calcium channel that plays a central role in calcium-dependent physiological responses essential for successful fertilization, such as sperm hyperactivation, acrosome reaction and chemotaxis towards the oocyte.
Specifications
CATSPER4 | |
Polyclonal | |
Unconjugated | |
CATSPER4 | |
4933412L05; cation channel of sperm 4; cation channel sperm associated 4; cation channel sperm-associated protein 4; cation channel, sperm associated 4; CatSper4; channel, sperm associated 4; CTSR4; ion channel CatSper4; RGD1565213 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
378807 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7RTX7 | |
CATSPER4 | |
Recombinant protein corresponding to Human CATSPER4. Recombinant protein control fragment (Product #RP-99995). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction