Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CATSPERB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557170
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557170 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557170 Supplier Invitrogen™ Supplier No. PA557170
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ILWLVDIPRE NITQSTDIAA VEEWLVRITL HHGLNIYATE GTLLDVIREP ILQWTPGDVI PESEISKLY Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 25%.

CATSPERB, also named as C14orf161 and CatSper-beta, is involved in sperm cell hyperactivation via its association with CATSPER1. CATSPERB has two isoforms with MW 127 kDa and 72 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CATSPERB
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CATSPERB
Gene Accession No. Q9H7T0
Gene Alias C14orf161; cation channel sperm associated auxiliary subunit beta; cation channel sperm-associated protein subunit beta; cation channel, sperm associated beta; cation channel, sperm-associated, beta; catsper channel auxiliary subunit beta; CatSper(beta); CATSPERB; CatSper-beta
Gene Symbols CATSPERB
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CATSPERB. Recombinant protein control fragment (Product #RP-94986).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79820
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.