Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CATSPERB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103718
Description
Antibody detects endogenous levels of total CATSPERB.
CATSPERB, also named as C14orf161 and CatSper-beta, is involved in sperm cell hyperactivation via its association with CATSPER1. CATSPERB has two isoforms with MW 127 kDa and 72 kDa.
Specifications
CATSPERB | |
Polyclonal | |
Unconjugated | |
CATSPERB | |
C14orf161; cation channel sperm associated auxiliary subunit beta; cation channel sperm-associated protein subunit beta; cation channel, sperm associated beta; cation channel, sperm-associated, beta; catsper channel auxiliary subunit beta; CatSper(beta); CATSPERB; CatSper-beta | |
Rabbit | |
Affinity chromatography | |
RUO | |
271036, 79820 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
A2RTF1, Q9H7T0 | |
CATSPERB | |
A synthesized peptide derived from human CATSPERB(Accession Q9H7T0), corresponding to amino acid residues M1077-E1116. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction