Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CATSPERD Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570119
Description
This target displays homology in the following species: Human: 100%.
Auxiliary component of the CatSper complex, a complex involved in sperm cell hyperactivation. Sperm cell hyperactivation is needed for sperm motility which is essential late in the preparation of sperm for fertilization. Required for CATSPER1 stability before intraflagellar transport and/or incorporation of the CatSper complex channel into the flagellar membrane.
Specifications
CATSPERD | |
Polyclonal | |
Unconjugated | |
CATSPERD | |
cation channel sperm associated auxiliary subunit delta; cation channel sperm-associated protein subunit delta; catsper channel auxiliary subunit delta; CATSPERD; catSperdelta; catSper-delta; TMEM146; Transmembrane protein 146 | |
Rabbit | |
Affinity chromatography | |
RUO | |
257062 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86XM0 | |
CATSPERD | |
synthetic peptide directed towards the N terminal region of human CATSPERD. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction