Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CATSPERD Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554968
Description
Immunogen sequence: RTDSNFTSSL KKATMSTLTV DIANKEISCV DIKPLSTLIS VGCDLDKKIV IQNKVSACSM GILDPLTLQD NYSFIIEKEF YDPGFQGQQS SEDLHVFYSY QQLGCPLLVY YDTLWKPVV Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 62%.
Auxiliary component of the CatSper complex, a complex involved in sperm cell hyperactivation. Sperm cell hyperactivation is needed for sperm motility which is essential late in the preparation of sperm for fertilization. Required for CATSPER1 stability before intraflagellar transport and/or incorporation of the CatSper complex channel into the flagellar membrane.
Specifications
CATSPERD | |
Polyclonal | |
Unconjugated | |
CATSPERD | |
cation channel sperm associated auxiliary subunit delta; cation channel sperm-associated protein subunit delta; catsper channel auxiliary subunit delta; CATSPERD; catSperdelta; catSper-delta; TMEM146; Transmembrane protein 146 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
257062 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86XM0 | |
CATSPERD | |
Recombinant protein corresponding to Human CATSPERD. Recombinant protein control fragment (Product #RP-90505). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction