Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CATSPERG Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543388
Description
Peptide sequence: FFLIQDLVTG DSGSFQGSYV LLVVGGGPTL DSLKDYSEDE IYRFNSPLDK Sequence homology: Guinea Pig: 86%; Human: 100%; Rat: 79%.
C19orf15 is a single-pass type I membrane protein. The exact function of C19orf15 remains unknown.
Specifications
| CATSPERG | |
| Polyclonal | |
| Unconjugated | |
| Catsperg | |
| C19orf15; cation channel sperm associated auxiliary subunit gamma; cation channel sperm-associated protein subunit gamma; cation channel, sperm-associated, gamma 1; catsper channel auxiliary subunit gamma; Catsperg; Catsperg1; RGD1310942 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 57828 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6ZRH7 | |
| Catsperg | |
| Synthetic peptide directed towards the C-terminal of human C19orf15 (aa 928-977). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction