Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Cav1.1 alpha1s, Mouse, Unlabeled, Clone: 1A,

Catalog No. MAB427CMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MAB427CMI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB427CMI Supplier MilliporeSigma™ Supplier No. MAB427C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Insulin receptor substrate 1 (UniProt P35568; also known as IRS-1) is encoded by the IRS1 gene (Gene ID 3667) in human. The insulin receptor substrates (IRS-1, IRS-2, IRS-3, and IRS-4) are adaptor proteins involved in modulating cell growth, metabolism, survival, and differentiation by transducing receptor signaling to multiple downstream effectors. IRS family proteins contain a conserved N-terminal pleckstrin homology (PH) domain (a.a. 12-115 of human IRS-1), a phosphotyrosine-binding (PTB) domain (a.a. 160-264 of human IRS-1), and multiple tyrosine phosphorylation sites in the C-terminal region, including Tyr612, Tyr896, Tyr941, Tyr1158, and Tyr1220. The PTB domain meidates the interaction of IRS-1, -2 and -3 with IR juxtamembrane NPXYp motif, facilitating subsequent receptor-mediated IRS tyrosine phosphorylation. Upon phosphorylation, IRS proteins serve as binding sites for Src homology 2 (SH2) domain-containing proteins, such as SHP-2 and PI 3-kinase. Functions of IRS proteins are largely dependent on the phosphorylations of specific tyrosine residues, which in turn are subject to negative regulation by phosphatase activity. In addition, IRS proteins can be phosphorylated by serine/threonine kinases, such as PI-3 kinase, Akt/PKB, GSK-3, mTOR, and PKC, and increased Ser/Thr phosphorylation reduces their binding to IR and tyrosine phosphorylation. Inhibition of PI 3-kinase enhances insulin-stimulated IRS-1 Tyr612/Tyr941 phosphorylation (p85-binding sites) and increases IRS-1 association with PI 3-kinase p85 subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cav1.1 alpha1s
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1A
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing PBS without preservatives.
Gene CACNA1S, CACH1, CACN1, CACNL1A3
Gene Accession No. P07293
Host Species Mouse
Immunogen Dihydropyridine-binding complex isolated from rabbit muscle t-tubules.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001095190
Test Specificity Clone 1A targets the extracellular region of voltage-dependent L-type calcium channel subunit alpha-1S or Cav1.1 encoded by the CACNA1S gene.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.