Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cav1.2 Antibody (S57), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP122439AF350
Description
Cav1.2 Monoclonal antibody specifically detects Cav1.2 in Human, Mouse, Rat, Hamster, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Cav1.2 | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Hamster, Rabbit | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
S57 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
alpha-1 subunit, calcium channel, voltage-dependent, L type, alpha 1C subunit, cardic dihydropyridine-sensitive, alpha-1 subunit, L type, alpha 1 polypeptide, isoform 1, cardic muscle, L type, alpha-1 polypeptide, isoform 1, cardiac muscle, splice variant 10* | |
Fusion protein amino acids 1507-1733 (intracellular carboxyl terminus) of rabbit Cav1.2 | |
0.1 mL | |
Cognition and Behavior, Phospho Specific | |
775 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction