Learn More
CaV1.2 Polyclonal Antibody, Invitrogen™
Guinea Pig Polyclonal Antibody
Supplier: Thermo Scientific PA5111791
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: (C)TTKINMD DLQPSENEDK S Homology: Mouse amino acid residues identical; guinea pig - 17/18 amino acid residues identical; human, rabbit - 16/18 amino acid residues identical.
CACNA1C encodes an alpha-1 subunit of a voltage-dependent calcium channel. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization. The alpha-1 subunit consists of 24 transmembrane segments and forms the pore through which ions pass into the cell. The calcium channel consists of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. There are multiple isoforms of each of these proteins, either encoded by different genes or the result of alternative splicing of transcripts. The protein encoded by CACNA1C binds to and is inhibited by dihydropyridine. Alternative splicing results in many transcript variants encoding different proteins.
Specifications
CaV1.2 | |
Polyclonal | |
Unconjugated | |
CACNA1C | |
CACB-receptor; Cach2; CACH3; Cacn2; CACN4; CACNA 1D; CACNA1C; Cacnl1a1; CACNL1A2; Calcium channel, L type, alpha-1 polypeptide, isoform 1, cardiac muscle; CaV1.2; CCHL1A1; D930026N18Rik; DHP receptor; I79_006562; long-NT; LQT8; LTCC; MBC; MELC-CC; MGC120730; Mouse brain class C; Rat brain class C; RATIVS302; RBC; Smooth muscle calcium channel blocker receptor; TS; Voltage-dependent L-type calcium channel subunit alpha-1C; Voltage-gated calcium channel subunit alpha Cav1.2 | |
Guinea Pig | |
Antigen affinity chromatography | |
RUO | |
12288, 24239, 775 | |
-20°C | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
P22002, Q01815, Q13936 | |
CACNA1C | |
Peptide corresponding to amino acid residues 848-865 of rat CaV1.2 (Intracellular loop between domains II and III) | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.