Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cav1.3 Antibody (S48), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212893AF647
Description
Cav1.3 Monoclonal antibody specifically detects Cav1.3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Cav1.3 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
S48 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
CACN4calcium channel, neuroendocrine/brain-type, alpha 1 subunit, CACNL1A2voltage-dependent L-type calcium channel subunit alpha-1D, Calcium channel, L type, alpha-1 polypeptide, isoform 2, calcium channel, voltage-dependent, L type, alpha 1D subunit, Cav1.3, CCHL1A2voltage-gated calcium channel alpha 1 subunit, L type, alpha-1 polypeptide, voltage-gated calcium channel alpha subunit Cav1.3, Voltage-gated calcium channel subunit alpha Cav1.3 | |
Fusion protein amino acids 859-875 of rat Cav1.3 | |
0.1 mL | |
Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
776 | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction