Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cav1.3 Antibody (S48), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212893
Description
Cav1.3 Monoclonal specifically detects Cav1.3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Microarray.Specifications
Cav1.3 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1 - 10 ug/mL, Immunohistochemistry 0.1 - 1 ug/mL, Immunocytochemistry/Immunofluorescence 1 - 10 ug/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin, Microarray | |
CACNA1D | |
Fusion protein amino acids 859-875 of rat Cav1.3 | |
0.1 mg | |
Primary | |
Detects approx 250 kDa. No cross-reactivity against Cav1.2. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2a κ |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay | |
S48 | |
Unconjugated | |
CACN4calcium channel, neuroendocrine/brain-type, alpha 1 subunit, CACNL1A2voltage-dependent L-type calcium channel subunit alpha-1D, Calcium channel, L type, alpha-1 polypeptide, isoform 2, calcium channel, voltage-dependent, L type, alpha 1D subunit, Cav1.3, CCHL1A2voltage-gated calcium channel alpha 1 subunit, L type, alpha-1 polypeptide, voltage-gated calcium channel alpha subunit Cav1.3, Voltage-gated calcium channel subunit alpha Cav1.3 | |
Mouse | |
Protein G purified | |
RUO | |
776 | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction