Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CaV1.3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120487
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120487 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120487 Supplier Invitrogen™ Supplier No. PA5120487
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Rat kidney. The target is usually found in the following locations: Membrane, Multi-pass membrane protein. Immunogen sequence: RQNYGYYSRY PGRNIDSERP RGYHHPQGFL EDDDSPVCYD SRRSPRRRLL PPTPASHRRS SFNFECLRRQ SSQEEVPSSP IFPHRTALPL HLMQQQIMAV AGLDSSKAQK YSPSHSTRSW ATPPATPPYR DWTPCYTPLI QVEQSEALDQ VNGSLPSLHR SSWYTDEPDI SYRTFTPASL TVPSSFRNKN SDKQRSADSL VEAVLISEGL GRYARDPKFV SATKHEIADA CDLTIDEMES AASTLLNGNV RPRANGDVGP LSHRQDYELQ DFGPGYSDEE PDPGRDEEDL ADEMICITTL.

CACNA1D is also known as PASNA, SANDD or Cav1. 3. Voltage-dependent calcium channels mediate the entry of calcium ions into excitable cells, and are also involved in a variety of calcium-dependent processes, including muscle contraction, hormone or neurotransmitter release, and gene expression. Calcium channels are multisubunit complexes composed of alpha-1, beta, alpha-2/delta, and gamma subunits. The channel activity is directed by the pore-forming alpha-1 subunit, whereas the others act as auxiliary subunits regulating this activity. The distinctive properties of the calcium channel types are related primarily to the expression of a variety of alpha-1 isoforms, namely alpha-1A, B, C, D, E, and S. This gene encodes the alpha-1D subunit. Several transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CaV1.3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.82 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CACNA1D
Gene Accession No. P27732, Q01668
Gene Alias 8430418G19Rik; brain class D; C79217; Cach3; Cacn4; CACNA 1D; CACNA1D; Cacnl1a2; calcium channel alpha-1 subunit; Calcium channel, L type, alpha-1 polypeptide isoform 2; Calcium channel, L type, alpha-1 polypeptide, isoform 2; calcium channel, neuroendocrine/brain-type, alpha 1 subunit; calcium channel, voltage-dependent, L type, alpha 1D subunit; calcium voltage-gated channel subunit alpha1 D; Cav1.3; CaV1.3alpha1; Cchl1a; CCHL1A2; D-LTCC; PASNA; Rat brain class D; RBD; SANDD; voltage-dependent calcium channel subunit alpha1D; voltage-dependent L-type calcium channel subunit alpha-1D; voltage-gated calcium channel alpha 1 subunit; voltage-gated calcium channel alpha subunit Cav1.3; voltage-gated calcium channel pore forming subunit CaV1.3alpha1 IVS3-IVS4 extracellular linker; voltage-gated calcium channel subunit alpha Cav1.3
Gene Symbols CACNA1D
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1882-2181 of human CACNA1D (NP_000711.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29716, 776
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.