Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAV1 Mouse anti-Human, Clone: Z034, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB14009
Description
Mouse monoclonal antibody raised against CAV1.
- High specificity
- High reactivity
Specifications
CAV1 | |
Monoclonal | |
Unconjugated | |
Immunofluorescence (2-4 ug/mL) Immunoprecipitation (2-5 ug/mL) Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user. | |
Q03135; A0M8T8 | |
CAV1 | |
Native purified tyrosine-phosphorylated CAV1 from RSV-transformed chick embryo fibroblasts. | |
100 μg | |
Primary | |
Human | |
Liquid |
Immunofluorescence, Immunoprecipitation, Western Blot | |
Z034 | |
Mouse monoclonal antibody raised against CAV1. | |
In PBS, pH 7.4 (0.09% sodium azide). | |
BSCL3/CAV/CGL3/MSTP085/VIP21 | |
Mouse | |
Affinity purification | |
RUO | |
857/373996 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction