Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CaV2.1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519598
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519598 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519598 Supplier Invitrogen™ Supplier No. PA519598
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

For Western Blot, this antibody has non-specific bands at 42 kDa. This antibody is predicted to react with rabbit, cow and human based on sequence homology. Store antibody at 4ºC for 1-2 weeks. For long-term storage, store at -20ºC.

Voltage-dependent calcium channels mediate the entry of calcium ions into excitable cells, and are also involved in a variety of calcium-dependent processes, including muscle contraction, hormone or neurotransmitter release, and gene expression. Calcium channels are multisubunit complexes composed of alpha-1, beta, alpha-2/delta, and gamma subunits. The channel activity is directed by the pore-forming alpha-1 subunit, whereas, the others act as auxiliary subunits regulating this activity. The distinctive properties of the calcium channel types are related primarily to the expression of a variety of alpha-1 isoforms, alpha-1A, B, C, D, E, and S. This gene encodes the alpha-1A subunit, which is predominantly expressed in neuronal tissue. Mutations in this gene are associated with 2 neurologic disorders, familial hemiplegic migraine and episodic ataxia 2. This gene also exhibits polymorphic variation due to n-repeats. Multiple transcript variants encoding different isoforms have been found for this gene. In one set of transcript variants, the n-repeats occur in the 3 UTR, and are not associated with any disease. But in another set of variants, an insertion extends the coding region to include the n-repeats which encode a polyglutamine tract. Expansion of the n-repeats from the normal 4-16 to 21-28 in the coding region is associated with spinocerebellar ataxia 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CaV2.1
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 98.98% PBS with 1% BSA and 0.02% sodium azide; pH 7.4
Gene CACNA1A
Gene Accession No. P54282, P97445
Gene Alias alpha1A; APCA; BccA1; BI; brain calcium channel 1; brain calcium channel I; brain class A; Caca1a; Cach4; CACN3; CACNA1A; Cacnl1a4; calcium channel alpha 1A; calcium channel, L type, alpha-1 polypeptide; Calcium channel, L type, alpha-1 polypeptide isoform 4; Calcium channel, L type, alpha-1 polypeptide, isoform 4; calcium channel, voltage-dependent, alpha 1A subunit; calcium channel, voltage-dependent, P/Q type, alpha 1A subunit; calcium voltage-gated channel subunit alpha1 A; Cav2.1; Ccha1a; EA2; fetal brain Ca2+ voltage-gated channel alpha1A pore-forming subunit; FHM; HPCA; la; leaner; MHP; MHP1; nmf352; Rat brain class A; rbA-1; RBA-I; rkr; rocker; SCA6; tg; tottering; voltage-dependent P/Q-type calcium channel subunit alpha-1A; Voltage-gated calcium channel subunit alpha Cav2.1
Gene Symbols CACNA1A
Host Species Rabbit
Immunogen Synthetic peptide conjugated to KLH derived from within residues 1 - 100 of Rat CACNA1A.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12286, 25398
Target Species Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.