Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CACNA1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109473
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109473 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109473 Supplier Invitrogen™ Supplier No. PA5109473
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SITYKTANSS PIHFAGAQTS LPAFSPGRLS RGLSEHNALL QRDPLSQPLA PGSRIGSDPY LGQRLDSEAS VHALPEDTLT FEEAVATNSG RSSRTSYVSS LTSQSHPLRR VPNGYHCTLG LSSGGRARHS YHHPDQDHWC.

The expression of multiple classes of voltage dependent calcium channels (VDCCs) allows neurons to tailor calcium signaling to functionally discrete cellular regions. Although N-Type VDCCs exist before birth which is consistent with a role in migration, most N-Type VDCCs subunit expression is postnatal and is important in the genesis of synaptic transmission in discrete hippocampal fields.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CACNA1B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CACNA1B
Gene Accession No. Q00975
Gene Alias alpha(1B); AW050276; AW060892; AW822256; BIII; Brain calcium channel III; Cach5; Cacna1b; Cacnl1a5; CACNN; calcium channel alpha12.2 subunit; calcium channel, L type, alpha-1 polypeptide; Calcium channel, L type, alpha-1 polypeptide isoform 5; calcium channel, voltage-dependent, L type, alpha 1B subunit; calcium channel, voltage-dependent, N type, alpha 1B subunit; calcium voltage-gated channel subunit alpha1 B; Cav2.2; Cav2.2 voltage-gated Ca2+ channel; Cchn1a; DYT23; voltage gated N-type calcium channel Ca(v)2.2; voltage-dependent N-type calcium channel subunit alpha-1B; voltage-gated calcium channel alpha subunit Cav2.2; voltage-gated calcium channel subunit alpha Cav2.2
Gene Symbols CACNA1B
Host Species Rabbit
Immunogen Synthetic peptide
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 774
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.