Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cav2.2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$219.50 - $487.50
Specifications
Antigen | Cav2.2 |
---|---|
Dilution | Western Blot 5 - 10 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Cav2.2 Polyclonal antibody specifically detects Cav2.2 in Human samples. It is validated for Western BlotSpecifications
Cav2.2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
BIII, Brain calcium channel III, CACNL1A5calcium channel, L type, alpha-1 polypeptide, CACNN, calcium channel alpha12.2 subunit, calcium channel, N type, calcium channel, voltage-dependent, alpha 1B subunit, N type, calcium channel, voltage-dependent, L type, alpha 1B subunit, calcium channel, voltage-dependent, N type, alpha 1B subunit, Cav2.2, Cav2.2 voltage-gated Ca2+ channel, L type, alpha-1 polypeptide isoform 5, voltage-dependent N-type calcium channel subunit alpha-1B, Voltage-gated calcium channel subunit alpha Cav2.2 | |
Synthetic peptide derived from the rat alpha1B calcium channel conjugated to KLH (Uniprot: Q00975) | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot 5 - 10 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS | |
774 | |
IgG | |
Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title