Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CaV2.3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109296
Description
Positive Control: Rat brain tissue, human kidney tissue, human placenta tissue, mouse brain tissue, 293T.
The expression of multiple classes of voltage dependent calcium channels (VDCCs) allows neurons to tailor calcium signaling to functionally discrete cellular regions. Although N-Type VDCCs exist before birth which is consistent with a role in migration, most N-Type VDCCs subunit expression is postnatal and is important in the genesis of synaptic transmission in discrete hippocampal fields.
Specifications
CaV2.3 | |
Polyclonal | |
Unconjugated | |
Cacna1e | |
A430040I15; alpha1E; BII; brain calcium channel II; CACH6; CACHA1E; CACNA1E; Cacnl1a6; calcium channel, L type, alpha-1 polypeptide, isoform 6; calcium channel, R type, alpha-1 polypeptide, isoform 6; calcium channel, voltage-dependent, L type, alpha 1E subunit; calcium channel, voltage-dependent, R type, alpha 1E subunit; calcium voltage-gated channel subunit alpha1 E; Cav2.3; Cchra1; gm139; RBE2; RBE-II; voltage gated calcium channel alpha1E subunit; voltage-dependent calcium channel alpha 1E subunit; voltage-dependent calcium channel alpha1-E subunit; voltage-dependent R-type calcium channel subunit alpha-1E; voltage-gated calcium channel alpha subunit Cav2.3; voltage-gated calcium channel subunit alpha Cav2.3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
54234, 777 | |
-20°C, Avoid Freeze/Thaw Cycles, store in dark | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
Q07652, Q15878 | |
Cacna1e | |
Synthetic peptide within rat CaV23 aa 875-924. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction