Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CaV3.1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537236
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537236 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537236 Supplier Invitrogen™ Supplier No. PA537236
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 262 kDa. Purity is >95% by SDS-PAGE.

Voltage-dependent Ca2+ channels mediate Ca2+ entry into excitable cells in response to membrane depolarization, and they are involved in a variety of Ca2+-dependent processes, including muscle contraction, hormone or neurotransmitter release and gene expression. Calcium channels are highly diverse, multimeric complexes composed of an alpha-1 subunit, an intracellular beta subunit, a disulfide linked alpha-2/delta subunit and a transmembrane gamma subunit. Ca2+ currents are characterized on the basis of their biophysical and pharmacologic properties and include L-, N-, T-, P-, Q-, and R- types. T-type Ca2+ currents are activated and inactivated more rapidly and at more negative membrane potentials than other Ca2+ current types. T-type Ca2+ channels enhance odor sensitivity by lowering the threshold of spike generation in olfactory receptor cells (ORCs).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CaV3.1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene CACNA1G
Gene Accession No. O43497
Gene Alias [a]1G; a1G; alpha-1G; Ca(V)T.1; CACNA1G; calcium channel, voltage-dependent, T type, alpha 1G subunit; calcium voltage-gated channel subunit alpha1 G; CaV T1; Cav3 1; cav3 1c; Cav3.1; Cav3.1c; Cav3.1d; KIAA1123; MGC117234; mKIAA1123; NBR13; SCA42; voltage-dependent calcium channel alpha 1G subunit, isoform 11; voltage-dependent T-type calcium channel alpha 1G subunit; voltage-dependent T-type calcium channel subunit alpha-1G; Voltage-gated calcium channel subunit alpha Cav3.1
Gene Symbols CACNA1G
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues in human CACNA1G.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8913
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.