Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cav3.2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP241303 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP241303 0.1 mg
NB396411 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP241303 Supplier Novus Biologicals Supplier No. NBP241303
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cav3.2 Polyclonal specifically detects Cav3.2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.

Specifications

Antigen Cav3.2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 2 ug/ml, ELISA, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin 5 ug/ml
Formulation PBS with 0.02% Sodium Azide
Gene Alias CACNA1HB, calcium channel, voltage-dependent, T type, alpha 1H subunit, calcium channel, voltage-dependent, T type, alpha 1Hb subunit, Cav3.2, ECA6, EIG6, FLJ90484, Low-voltage-activated calcium channel alpha1 3.2 subunit, low-voltage-activated calcium channel alpha13.2 subunit, voltage dependent t-type calcium channel alpha-1H subunit, voltage-dependent T-type calcium channel subunit alpha-1H, voltage-gated calcium channel alpha subunit Cav3.2, voltage-gated calcium channel alpha subunit CavT.2, Voltage-gated calcium channel subunit alpha Cav3.2
Gene Symbols CACNA1H
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid peptide near the center of human CACNA1H. The immunogen is located within amino acids 1010 - 1060 of CACNA1H. Amino Acid Squence: AEGDANRSDTDEDKTS
Molecular Weight of Antigen 260 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8912
Test Specificity Multiple isoforms of CACNA1H are known to exist.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.