Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CaV3.2 Polyclonal Antibody
CaV3.2 Polyclonal Antibody
Supplier: Thermo Scientific PA572836
Description
CaV3.2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Specifications
CaV3.2 | |
Polyclonal | |
Unconjugated | |
CACNA1H | |
A1H-b; alpha13.2; CACNA1H; CACNA1HB; calcium channel alpha13.2 subunit; calcium channel, voltage-dependent, T type, alpha 1H subunit; calcium channel, voltage-dependent, T type, alpha 1Hb subunit; calcium voltage-gated channel subunit alpha1 H; Cav3.2; ECA6; EIG6; FLJ90484; Low-voltage-activated calcium channel alpha1 3.2 subunit; low-voltage-activated calcium channel alpha13.2 subunit; MNCb-1209; T-type Cav3.2; voltage dependent t-type calcium channel alpha-1H subunit; voltage-dependent T-type calcium channel subunit alpha-1H; voltage-gated calcium channel alpha subunit Cav3.2; voltage-gated calcium channel alpha subunit CavT.2; voltage-gated calcium channel subunit alpha Cav3.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114862, 58226, 8912 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O88427, O95180, Q9EQ60 | |
CACNA1H | |
CACNA1H antibody for a 16 amino acid peptide near the center of human CACNA1H. The immunogen is within amino acids 1010 - 1060 of CACNA1H. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction