Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caveolin-1 Antibody (M281), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP323193
Description
Caveolin-1 Monoclonal antibody specifically detects Caveolin-1 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Caveolin-1 | |
Monoclonal | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Chicken | |
Purified |
Western Blot, Immunofluorescence | |
M281 | |
Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:50 | |
CAV, caveolae protein, 22kD, caveolin 1, caveolae protein, 22kDa, cell growth-inhibiting protein 32, MSTP085 | |
Clone (M281) was generated from a full length recombinant human Caveolin-1 protein. [UniProt# Q03135] | |
0.1 mL | |
Cancer, Caveolae Markers, Cellular Markers, Cholesterol Metabolism, Hypoxia, Lipid and Metabolism, Lipid Droplets, Phospho Specific, Signal Transduction | |
857 | |
Store at -20°C. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction