Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caveolin-1 Monoclonal Antibody (A3E8), Huabio

Catalog No. 502544790
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4790 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4790 Supplier HUABIO RESEARCH INC Supplier No. EM190188
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Caveolin-1 Monoclonal antibody specifically detects Caveolin-1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Caveolin-1
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone A3E8
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene CAV1
Gene Accession No. Q03135
Gene Alias BSCL3; CAV; CAV1; Cav-1; caveolae; caveolin 1; caveolin 1, caveolae protein; caveolin 1, caveolae protein, 22kDa; caveolin 1a; caveolin 1b; caveolin caveolae protein 22 kDa; caveolin, caveolae protein 1; caveolin, caveolae protein, 22 kDa; caveolin-1; caveolin-1a; caveolin-1b; cell growth-inhibiting protein 32; CGL3; HGNC:1527; LCCNS; MSTP085; OTTHUMP00000196475; PPH3; Unknown (protein for IMAGE:7985573); variant a; variant b; vesicular integral-membrane protein VIP21; VIP21; wu:fc07c04
Host Species Mouse
Immunogen Recombinant protein within human Caveolin-1 aa 1-150.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 857
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.