Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caveolin-2 Goat anti-Human, Mouse, Rat, Alexa Fluor 488, Clone: , R&D Systems™

Catalog No. IC5788G
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC5788G 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC5788G Supplier R&D Systems Supplier No. IC5788G
This item is not returnable. View return policy

Goat Polyclonal Antibody

Caveolin-2 is a cell membrane-associated member of the caveolin family of proteins. Although its predicted MW is 17kDa, it runs anomalously at 21-22kDa on SDS-PAGE. Caveolin-2 is not ubiquitous, but it is found in divergent cell types such as smooth muscle, endothelial, and skeletal muscle cells, plus fibroblasts and adipocytes. When expressed alone, Caveolin-2 forms monomers and homodimers; when expressed with Caveolin-1, it exists as part of a 150kDa, Caveolin-1 associated oligomeric complex. Human Caveolin-2 is 162 amino acids (aa) in length. It contains a central membrane-embedded sequence (aa 87-107) flanked by two cytoplasmic domains (aa 1-86 and 108-162). Caveolin-2 has been found phosphorylated at tyrosines 19 and 27, and at serines 20, 23 and 36.

Specifications

Antigen Caveolin-2
Applications Immunocytochemistry
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Immunocytochemistry 1:10 dilution
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P51636
Gene Alias CAV, CAV2, caveolae protein, 20-kD, caveolin 2, caveolin 2 isoform a and b, caveolin-2, Caveolin2, MGC12294
Gene Symbols CAV2
Host Species Goat
Immunogen E. coli-derived recombinant human Caveolin-2, Met1-Lys86, Accession # P51636
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 858
Test Specificity Caveolin-2 antibodies are ideal for immunocytochemistry colocalization studies in caveolae. The unconjugated antibody detects endogenous human, mouse, and rat Caveolin-2 in Western blots.
Target Species Human, Mouse, Rat
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.