Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caveolin-3 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298712MFV610
Description
Caveolin-3 Polyclonal antibody specifically detects Caveolin-3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Caveolin-3 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
caveolin 3, caveolin-3, LGMD1CMGC126100, LQT9MGC126101, M-caveolin, MGC126129, VIP21, VIP-21 | |
Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the Human Caveolin-3. | |
0.1 mL | |
Endocrinology, Signal Transduction | |
859 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction