Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caveolin-3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB1105029
Description
Caveolin-3 Polyclonal specifically detects Caveolin-3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
Caveolin-3 | |
Polyclonal | |
Unconjugated | |
PBS, 1 mg/ml BSA with 0.05% Sodium Azide | |
caveolin 3, caveolin-3, LGMD1CMGC126100, LQT9MGC126101, M-caveolin, MGC126129, VIP21, VIP-21 | |
Rabbit | |
Affinity Purified | |
RUO | |
859 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
1 mg/mL | |
Western Blot 1 - 3 ug/ml, Flow Cytometry 3 - 5 ug 10^6 cells, Immunohistochemistry 1:100 - 200, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunoprecipitation 5 ug, Immunohistochemistry-Paraffin 1:100 - 200 | |
P56539 | |
CAV3 | |
Synthetic peptide corresponding to residues M(1) M T E E H T D L E A R I I K D I H C(19) C of mouse CAV3 | |
100 μg | |
Primary | |
Detects caveolin-3 from rat tissues. This does not detect caveolin-1 or -2. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction