Learn More
Mopec™ Cavity Chemical Injector
Shop All Mopec Products
Click to view available options
Material:
Stainless Steel
Description
- Leakproof under all conditions
- Fits any standard threaded fluid bottle
Specifications
Specifications
Autoclavable | Not Autoclavable |
Material | Stainless Steel |
Length (English) | 6.25 in. |
Length (Metric) | 16 cm |