Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAZIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520912
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520912 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520912 Supplier Thermo Scientific Supplier No. PA520912
Add to Cart
Add to Cart

CAZIP Polyclonal Antibody

CAZIP Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen CAZIP
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene MTUS2
Gene Accession No. Q3UHD3, Q5JR59
Gene Alias +TIP of 150 kDa; 5730592G18Rik; A730013O20Rik; AU067634; C130038G02Rik; Cardiac zipper protein; CAZIP; Gm763; ICIS; Kiaa0774; microtubule associated tumor suppressor candidate 2; microtubule plus-end tracking protein TIP150; Microtubule-associated tumor suppressor candidate 2; microtubule-associated tumor suppressor candidate 2 homolog; mKIAA0774; Mtus2; RGD1562533; RP11-274A8.2; Tip150; tracking protein of 150 kDa
Gene Symbols MTUS2
Host Species Rabbit
Immunogen An 18 amino acid peptide near the carboxy terminus of human CAZIP.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23281, 77521
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.