Learn More
Description
CB-103 is a highly specific protein-protein interaction (PPI) inhibitor that interferes with the interaction between the active intracellular domains of Notch receptors (NICD) and the CSL transcription factor complex. In combination with ICI 182,780 (Cat. No. 1047), CB-103 arrests the growth of mouse xenografts. In mouse models, it displays potent tumor growth inhibition and delayed tumor relapse when combined with Taxol (Cat. No. 1097). CB-103 inhibits differentiation of mural cells by downregulating Notch pathway. CB-103 is orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 6-[4-(1,1-Dimethylethyl)phenoxy]-3-pyridinamine |
| CAS | 218457-67-1 |
| Quantity | 10 mg |
| Product Line | Tocris |
| Target | Notch Signaling Inhibitors |
| Molecular Formula | C15H18N2O |
| Purity | 98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
