Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CB 5083
GREENER_CHOICE

Catalog No. 737925
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
73-792-5 25 mg
73-795 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 73-792-5 Supplier Tocris Bioscience™ Supplier No. 7379/25
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent p97 ATPase inhibitor

CB 5083 is a potent p97 inhibitor, inhibiting the D2 ATPase domain (IC50 = 11 nM). CB 5083 shows broad antitumor activity and inhibits tumor growth in vivo. CB 5083 is cytotoxic to A549 cancer cells (IC50 = 680 nM), increases CHOP and GRP78 protein levels and results in the loss of aggresomes.

Specifications

Chemical Name or Material 1-[7,8-Dihydro-4-[(phenylmethyl)amino]-5 H-pyrano[4,3-d]pyrimidin-2-yl]-2-methyl-1 H-indole-4-carboxamide
CAS 1542705-92-9
Quantity 25 mg
Target p97 ATPase Inhibitors
Molecular Formula C24H23N5O2
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.