Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CB 839
GREENER_CHOICESDP

Catalog No. 759150 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-915-0 50 mg
75-911-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 75-915-0 Supplier Tocris Bioscience™ Supplier No. 7591/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent glutaminase GLS1 inhibitor

CB 839 is a potent non-competitive glutaminase (GLS1) inhibitor (IC50 = 24 nM for recombinant human GAC). The compound exhibits selectivity for GLS1 over GLS2. CB 839 displays antiproliferative activity in a triple-negative breast cancer cell line (GI50 values are 19 and 55 nM against MDA-MB-231 and HCC1806, respectively); it reduces glutamine consumption and glutamate production rates in HCC1806 cells. CB 839 displays significant antitumor activity as a single agent in a patient-derived TNBC xenografts model. CB 839 inhibits the growth of basal-like HER2 cell line JIMT-1 xenografts in mice, both as a single agent and in combination with Paclitaxel (Cat. No. 1097). CB 839 works with Erlotinib (Cat. No. 7194) to reduce glucose and glutamine uptake, increase energetic stress and activate the AMP-activated protein kinase (AMPK) pathway in EGFR (del19) non-small cell lung cancer xenografts in vivo. CB 839 is orally bioavailable.

Specifications

Chemical Name or Material N-[5-[4-[6-[[2-[3-(Trifluoromethoxy)phenyl]acetyl]amino]-3-pyridazinyl]butyl]-1,3,4-thiadiazol-2-yl]-2-pyridineacetamide
CAS 1439399-58-2
Quantity 50 mg
Target Glutaminase Inhibitors
Molecular Formula C26H24F3N7O3S
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.