Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBARA1, Rabbit anti-Human,Mouse,Rat, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP280412

Catalog No. NB013484


Description

Description

CBARA1 Polyclonal specifically detects CBARA1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

CBARA1
Polyclonal
Unconjugated
PBS (pH7.4), 0.2% BSA, 50% Glycerol with 0.05% Sodium Azide
MICU1
Recombinant protein within Human CBARA1 aa 14-154/ 476. (SwissProt: Q9BPX6 Human; SwissProt: Q8VCX5 Mouse; SwissProt: Q6P6Q9 Rat)
100 μL
Primary
Human, Mouse, Rat
IgG
Western Blot, Immunohistochemistry (Paraffin)
1 mg/mL
Western Blot 1:500-1:2000
Allergen Hom s 4, ara CALC, Atopy-related autoantigen CALC, CALCDKFZp564C246, calcium binding atopy-related autoantigen 1, Calcium-binding atopy-related autoantigen 1, MICU1mitochondrial, mitochondrial calcium uptake 1
Rabbit
Immunogen affinity purified
RUO
10367
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.