Learn More
Invitrogen™ CBF beta Polyclonal Antibody
Description
CBF beta Polyclonal Antibody for ICC/IF, IHC (P), Western Blot
Specifications
Specifications
| Antigen | CBF beta |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene | CBFB |
| Gene Accession No. | Q13951 |
| Gene Alias | AI893578; CBFB; CBF-beta; core binding factor beta; core-binding factor subunit beta; core-binding factor, beta subunit; PEA2; PEA2-beta; Pebp2; PEBP2B; PEBP2-beta; Pebpb2; polyomavirus enhancer binding protein 2, beta subunit; polyomavirus enhancer-binding protein 2 beta subunit; SL3/AKV core-binding factor beta subunit; SL3-3 enhancer factor 1 beta subunit; SL3-3 enhancer factor 1 subunit beta |
| Gene Symbols | CBFB |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.