Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBF1-interacting corepressor Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171896JF549
Description
CBF1-interacting corepressor Polyclonal antibody specifically detects CBF1-interacting corepressor in Human samples. It is validated for Western BlotSpecifications
CBF1-interacting corepressor | |
Polyclonal | |
Western Blot | |
CBF1 interacting corepressor, CBF1-interacting corepressor, CIRrecepin, corepressor interacting with RBPJ 1, corepressor interacting with RBPJ, 1, Recepin | |
The immunogen for this product maps to a region between residue 400 and 450 of human Corepressor Interacting with RBPJ 1 using the numbering given in entry NP_004873.3 (GeneID 9541). | |
0.1 mL | |
Cell Biology | |
9541 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction