Learn More
CBFA2T2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303593A

Description
The recommended shelf life for this product is 1 year from date of receipt.
In acute myeloid leukemia, especially in the M2 subtype, the t(8;21)(q22;q22) translocation is one of the most frequent karyotypic abnormalities. The translocation produces a chimeric gene made up of the 5′-region of the RUNX1 (AML1) gene fused to the 3′-region of the CBFA2T1 (MTG8) gene. The chimeric protein is thought to associate with the nuclear corepressor/histone deacetylase complex to block hematopoietic differentiation. The protein encoded by this gene binds to the AML1-MTG8 complex and may be important in promoting leukemogenesis. Several transcript variants are thought to exist for this gene, but the full-length natures of only three have been described.Specifications
CBFA2T2 | |
Polyclonal | |
Unconjugated | |
CBFA2T2 | |
DKFZp313F2116, EHT, ZMYND3 CBFA2T2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9139 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
O43439 | |
CBFA2T2 | |
Between 554 and 604 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.