Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBFA2T3 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204048JF646
Description
CBFA2T3 Polyclonal antibody specifically detects CBFA2T3 in Human samples. It is validated for Western BlotSpecifications
CBFA2T3 | |
Polyclonal | |
Western Blot | |
core-binding factor, runt domain, alpha subunit 2; translocated to, 3, hMTG16, MTG16ETO2, MTG8-related protein 2, MTGR2MTG8-related gene 2, Myeloid translocation gene on chromosome 16 protein, protein CBFA2T3, Zinc finger MYND domain-containing protein 4, ZMYND4myeloid translocation gene 8 and 16b | |
C-terminus: between aa 603 and 653 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
863 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction