Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBFA2T3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25763625UL
Description
CBFA2T3 Polyclonal specifically detects CBFA2T3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CBFA2T3 | |
Polyclonal | |
Western Blot 0.4 μg/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
core-binding factor, runt domain, alpha subunit 2; translocated to, 3, hMTG16, MTG16ETO2, MTG8-related protein 2, MTGR2MTG8-related gene 2, Myeloid translocation gene on chromosome 16 protein, protein CBFA2T3, Zinc finger MYND domain-containing protein 4, ZMYND4myeloid translocation gene 8 and 16b | |
Rabbit | |
Affinity Purified | |
RUO | |
863 | |
Human | |
IgG |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
CBFA2T3 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SRLRDRAASSASGSTCGSMSQTHPVLESGLLASAGCSAPRGPRKGGPAPVDRKAKASAMPDSPAEVKTQPR | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction