Learn More
CBFA2T3 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303616AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a member of the myeloid translocation gene family which interact with DNA-bound transcription factors and recruit a range of corepressors to facilitate transcriptional repression. The t(16;21)(q24;q22) translocation is one of the less common karyotypic abnormalities in acute myeloid leukemia. The translocation produces a chimeric gene made up of the 5′-region of the runt-related transcription factor 1 gene fused to the 3′-region of this gene. This gene is also a putative breast tumor suppressor. Alternative splicing results in transcript variants. [provided by RefSeq, Nov 2010].Specifications
CBFA2T3 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O75081 | |
CBFA2T3 | |
Between 603 and 653 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CBFA2T3 | |
Cbfa2t3h, Mtgr2, MTG16HEL | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
863 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.