Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBFB Antibody (PCRP-CBFB-1E6), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314059PCP
This item is not returnable.
View return policy
Description
CBFB Monoclonal antibody specifically detects CBFB in Human samples. It is validated for Flow CytometrySpecifications
| CBFB | |
| Monoclonal | |
| PerCP | |
| CBF-beta, core-binding factor subunit beta, core-binding factor, beta subunit, PEA2-beta, PEBP2B, PEBP2-beta, polyomavirus enhancer binding protein 2, beta subunit, Polyomavirus enhancer-binding protein 2 beta subunit, SL3/AKV core-binding factor beta subunit, SL3-3 enhancer factor 1 beta subunit, SL3-3 enhancer factor 1 subunit beta | |
| Recombinant full-length human CBFB protein (Uniprot: Q13951) | |
| 0.1 mL | |
| DNA Replication Transcription Translation and Splicing | |
| 865 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry | |
| PCRP-CBFB-1E6 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction